
2,3-Difluorobenzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4519-39-5
:
Formula:C₇H₄F₂O₂
MDL Number:MFCD00010267
:158.1 g/mol
Melting Pt:160…164 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together