

2,4-Dichloro-5-isopropoxyaniline ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:41200-96-8
:
Formula:C₉H₁₁Cl₂NO
MDL Number:MFCD00974411
:220.1 g/mol
Boiling Pt:110…112 °C (0.2 mmHg)
Melting Pt:40…44 °C
Storage Temperature:Ambient
EINECS:255-258-9
UN:2811
ADR:6.1,III
:GHS06
:GHS07
Related Information
Frequently Bought Together