

4-Chloro-3-fluorophenol ≥98%
Catalog # A16029.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
CAS Number:348-60-7
:
Formula:C₆H₄ClFO
MDL Number:MFCD00042583
:146.55 g/mol
Boiling Pt:84 °C (44 mmHg)
Melting Pt:50…54 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
:GHS07