

2,6-Dichloro-4-fluorophenol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:392-71-2
:
Formula:C₆H₃Cl₂FO
MDL Number:MFCD00010675
:180.99 g/mol
Boiling Pt:142 °C (100 mmHg)
Melting Pt:47…51 °C
Flash Pt:103 °C (217 °F)
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS06
:GHS05
Related Information
Frequently Bought Together