
5-Amino-2-chlorobenzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:89-54-3
:
Formula:C₇H₆ClNO₂
MDL Number:MFCD00007845
:171.58 g/mol
Boiling Pt:330 °C (1013 hPa)
Melting Pt:∼ 185 °C (dec.)
Density:1.5
Storage Temperature:Ambient
EINECS:201-916-5
:GHS07
Related Information
Frequently Bought Together