
3-Chlorobenzoic acid
Catalog # EHERC11390800
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:156.57 g/mol
Merck Index:13,02142
Boiling Pt:281 °C (1013 hPa)
Melting Pt:122 °C
Density:1.496 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-618-4
:GHS07
CAS Number:535-80-8
Formula:C₇H₅ClO₂
MDL Number:MFCD00002491