
2-Chlorobenzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:118-91-2
:
Formula:C₇H₅ClO₂
MDL Number:MFCD00002412
:156.57 g/mol
Merck Index:14,2124
Boiling Pt:284…286 °C
Melting Pt:138…142 °C
Density:1.544
Flash Pt:173 °C (343 °F)
EINECS:204-285-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together