Chlorobenzene 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-90-7
:
Formula:C₆H₅Cl
MDL Number:MFCD00000530
:112.56 g/mol
Merck Index:13,02139
Boiling Pt:131 °C (760 mmHg)
Melting Pt:–45 °C
Density:1.1075 g/cm³
Flash Pt:23 °C
Storage Temperature:Ambient
EINECS:203-628-5
UN:1134
ADR:3,III
REACH:01-2119432722-45
:GHS07
:GHS09
:GHS02
Specification Test Results
- Appearance:Clear liquid
- GC:≥99.0 %
- UV:at 288 nm A: ≤1.0
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0005 %
Specification Test Results
Frequently Bought Together