Order Entry
Czech Republic
ContactUsLinkComponent
1,4-Dichlorobenzene
Health
Environment
1,4-Dichlorobenzene
Catalog # EHERC12372000
Supplier:  EHRENSTORFER
CAS Number:  
Health
Environment
1,4-Dichlorobenzene
Catalog # EHERC12372000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :147 g/mol
Merck Index:13,03082
Boiling Pt:173.4 °C (1013 hPa)
Melting Pt:48…50 °C
Density:1.458 g/cm³ (20 °C)
Flash Pt:65 °C
Storage Temperature:Ambient
EINECS:203-400-5
UN:3077
ADR:9,III
 :GHS08
 :GHS09
CAS Number:106-46-7
Formula:C₆H₄Cl₂
MDL Number:MFCD00000604