Order Entry
Czech Republic
ContactUsLinkComponent
1,2-Dichlorobenzene for synthesis, Sigma-Aldrich®
Irritant
Environment
1,2-Dichlorobenzene for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
Environment
1,2-Dichlorobenzene for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    0.13 g/l (20 °C)
  • Melting Point:
    -17 °C
  • Molar Mass:
    147 g/mol
  • Boiling Point:
    180 °C (1013 hPa)
  • Vapor Pressure:
    1.33 hPa (20 °C)
  • Flash Point:
    66 °C
  • Explosion Limit:
    2.2 - 12 %(V)
  • Density:
    1.31 g/cm3 (20 °C)
  • Ignition Point:
    640 °C
  • Synonyms:
    o-Chlorobenzene
  • Hill Formula:
    C6H4Cl2
  • Chemical Formula:
    1,2-(Cl)2C6H4

Specification Test Results

Related Information

Frequently Bought Together