Order Entry
Czech Republic
ContactUsLinkComponent
2-Phenoxybenzoic acid
2-Phenoxybenzoic acid
Catalog # EHERC16045100
Supplier:  EHRENSTORFER
CAS Number:  
2-Phenoxybenzoic acid
Catalog # EHERC16045100
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :214.22 g/mol
Boiling Pt:355 °C (1013 hPa)
Melting Pt:113…115 °C
Density:1.2141 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:218-811-5
CAS Number:2243-42-7
Formula:C₁₃H₁₀O₃
MDL Number:MFCD00002429