
2,4-Dihydroxybenzoic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:89-86-1
:
Formula:C₇H₆O₄
MDL Number:MFCD00002451
Merck Index:12,08325
Boiling Pt:390 °C (1013 hPa)
Melting Pt:210…215 °C
Density:1,54 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-946-9
:GHS07
Specification Test Results
- Appearance:Cream to light beige to pink Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- Titration with NaOH:≥96.0 %
Specification Test Results
Frequently Bought Together