
2,6-Dimethoxybenzoic acid ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1466-76-8
:
Formula:C₉H₁₀O₄
MDL Number:MFCD00002437
:182.18 g/mol
Boiling Pt:315 °C (1013 hPa)
Melting Pt:∼188 °C (dec.)
Density:1.365 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-985-4
:GHS07
Related Information
Frequently Bought Together