Order Entry
Czech Republic
ContactUsLinkComponent
2,6-Dimethoxybenzoic acid 99%
Irritant
2,6-Dimethoxybenzoic acid 99%
Katalogové číslo ACRO115440250
CAS Number:  
undefined
Irritant
2,6-Dimethoxybenzoic acid 99%
Katalogové číslo ACRO115440250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    187 to 190 °C
  • Water:
    ≤0.5 % (K.F.)
  • HPLC:
    ≥98.5 %
  • Titration with NaOH:
    ≥98.5 %

Specification Test Results

About this item

 :
 :182.18 g/mol
Boiling Pt:315 °C (1013 hPa)
Melting Pt:187…190 °C
Density:1,365 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-985-4
 :GHS07
CAS Number:1466-76-8
Formula:C₉H₁₀O₄
MDL Number:MFCD00002437