
2,5-Dimethoxybenzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2785-98-0
:
Formula:C₉H₁₀O₄
MDL Number:MFCD00002436
Boiling Pt:314 °C (1013 hPa)
Melting Pt:75…79 °C
:182.18 g/mol
Storage Temperature:Ambient
EINECS:220-503-0
:GHS07
Related Information
Frequently Bought Together