Order Entry
Czech Republic
ContactUsLinkComponent
4-Nitrobenzoic acid for synthesis, Sigma-Aldrich®
Irritant
4-Nitrobenzoic acid for synthesis, Sigma-Aldrich®
Catalog # 8.00646.0100
Supplier:  Merck
CAS Number:  
Irritant
4-Nitrobenzoic acid for synthesis, Sigma-Aldrich®
Catalog # 8.00646.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    0.42 g/l (20 °C)
  • Melting Point:
    239 - 242 °C
  • Molar Mass:
    167.12 g/mol
  • Bulk Density:
    700 kg/m3
  • Density:
    1.61 g/cm3 (20 °C)
  • pH:
    3.1 (0.42 g/l, H2O, 20 °C) (saturated solution)
  • Synonyms:
    4-Nitrobenzoic acid
  • Hill Formula:
    C7H5NO4
  • Chemical Formula:
    4-(NO2)C6H4COOH

Specification Test Results

About this item

 :
 :167.12 g/mol
Merck Index:12,06686
Boiling Pt:359 °C (1013 hPa)
Melting Pt:121 °C
Density:1.65 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-526-2
 :GHS07
CAS Number:62-23-7
Formula:C₇H₅NO₄
MDL Number:MFCD00007352