
4-(Dimethylamino)benzoic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:619-84-1
:
Formula:C₉H₁₁NO₂
MDL Number:MFCD00002537
:165.19 g/mol
Merck Index:13,03259
Boiling Pt:297 °C (1013 hPa)
Melting Pt:238…242 °C
Density:1,29 g/cm³ (20 °C)
Flash Pt:170 °C
EINECS:210-615-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to beige to grey-green Crystalline powder
- Infrared spectrum:Conforms
- Melting point:238 to 242 °C
- Water:≤0.5 % (K.F.)
- Titration with NaOH:≥97.5 % (On dry substance)
Specification Test Results
Frequently Bought Together