3-(Dimethylamino)benzoic acid for synthesis, Sigma-Aldrich®
Catalog # 8.14112.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:149 - 152 °C
- Molar Mass:165.19 g/mol
- Hill Formula:C9H11NO2
- Chemical Formula:3-((CH3)2N)C6H4COOH
Specification Test Results
About this item
:
:165.19 g/mol
Boiling Pt:298 °C (1013 hPa)
Melting Pt:149…152 °C
Density:1.26 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:202-775-2
CAS Number:99-64-9
Formula:C₉H₁₁NO₂
MDL Number:MFCD00002497