Order Entry
Czech Republic
ContactUsLinkComponent
3-(Dimethylamino)benzoic acid 98%
3-(Dimethylamino)benzoic acid 98%
Catalog # A13612.14
CAS Number:  
3-(Dimethylamino)benzoic acid 98%
Catalog # A13612.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :165.19 g/mol
Boiling Pt:298 °C (1013 hPa)
Melting Pt:148…153 °C
Density:1.26 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-775-2
CAS Number:99-64-9
Formula:C₉H₁₁NO₂
MDL Number:MFCD00002497