Order Entry
Czech Republic
ContactUsLinkComponent
3-(Dimethylamino)benzoic acid 99%
Irritant
3-(Dimethylamino)benzoic acid 99%
Certificates
undefined
Irritant
3-(Dimethylamino)benzoic acid 99%

Specification Test Results

  • Appearance:
    Light yellow to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    150 to 153 °C
  • Titration with NaOH:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together