Order Entry
Czech Republic
ContactUsLinkComponent
Benzoic acid, Supelco®
Benzoic acid, Supelco®
Catalog # 73983-5G-F
Supplier:  Merck
CAS Number:  
undefined
Benzoic acid, Supelco®
Catalog # 73983-5G-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :122.12 g/mol
Merck Index:13,01092
Boiling Pt:249 °C (1013 hPa)
Melting Pt:121…123 °C
Density:1.316 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Flash Pt:121 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-618-2
REACH:01-2119455536-33
 :GHS07
CAS Number:65-85-0
Formula:C₇H₆O₂
MDL Number:MFCD00002398