Order Entry
Czech Republic
ContactUsLinkComponent
Benzoic acid for synthesis, Sigma-Aldrich®
Corrosive
Health
Benzoic acid for synthesis, Sigma-Aldrich®
Catalog # 8.22257.0100
Supplier:  Merck
CAS Number:  
undefined
Corrosive
Health
Benzoic acid for synthesis, Sigma-Aldrich®
Catalog # 8.22257.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    2.9 g/l (25 °C)
  • Melting Point:
    121 - 123 °C
  • Molar Mass:
    122.12 g/mol
  • Bulk Density:
    500 kg/m3
  • Boiling Point:
    249 °C (1013 hPa)
  • Vapor Pressure:
    0.001 hPa (20 °C)
  • Flash Point:
    121 °C
  • Density:
    1.321 g/cm3 (20 °C)
  • pH:
    3.1 (1 g/l, H2O, 20 °C)
  • Ignition Point:
    570 °C

Specification Test Results

About this item

 :
 :122.12 g/mol
Merck Index:13,01092
Boiling Pt:249 °C (1013 hPa)
Melting Pt:121…123 °C
Density:1.316 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Flash Pt:121 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-618-2
REACH:01-2119455536-33
 :GHS05
 :GHS08
CAS Number:65-85-0
Formula:C₇H₆O₂
MDL Number:MFCD00002398