Order Entry
Czech Republic
ContactUsLinkComponent
 
Benzoic acid, Supelco®
  1.00134.0005
 :  Merck
 :  
undefined
Benzoic acid, Supelco®
  1.00134.0005
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Solubility:
    2.9 g/l (25 °C)
  • Melting Point:
    121 - 123 °C
  • Molar Mass:
    122.12 g/mol
  • Bulk Density:
    500 kg/m3
  • Boiling Point:
    249 °C (1013 hPa)
  • Vapor Pressure:
    0.001 hPa (20 °C)
  • Flash Point:
    121 °C
  • Density:
    1.321 g/cm3 (20 °C)
  • pH:
    2.5 - 3.5 (H2O, 20 °C) (saturated solution)
  • Ignition Point:
    570 °C

 

 

 :
 :122.12 g/mol
 :13,01092
 :249 °C (1013 hPa)
 :121…123 °C
 :1.316 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
 :121 °C (closed cup)
 :Ambient
 :200-618-2
 :01-2119455536-33
 :GHS05
 :GHS08
 :65-85-0
 :C₇H₆O₂
 :MFCD00002398