Benzoic acid 99.5% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:65-85-0
:
Formula:C₇H₆O₂
MDL Number:MFCD00002398
:122.12 g/mol
Merck Index:13,01092
Boiling Pt:249 °C (760 mmHg)
Melting Pt:121…123 °C
Density:1,316 g/cm³ (20 °C)
Flash Pt:121 °C
Storage Temperature:Ambient
EINECS:200-618-2
REACH:01-2119455536-33
:GHS05
:GHS08
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:121 to 123 °C
- Sulfated ash:≤0.01 %
- Titration with NaOH:≥99.5 %
- Sulfate (SO4):≤30 ppm
- Iron (Fe):≤4 ppm
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.005 % (in chloroform)
Specification Test Results
Frequently Bought Together