Burettes, straight stopcock, Schellbach stripe, DURAN®,
Supplier: DWK Life Sciences
Certificates
About this item
DURAN®, borosilicate glass 3.3, clear, class AS.
- With Schellbach stripe and main graduations as circular divisions
- Straight stopcock
- With glass key
- Marks and inscription in blue
- Calibration is based on the poured out volume ("Ex")
Complies to the DIN ISO 385
With printed retrace code (batch identification).
Frequently Bought Together