p-Anisaldehyde 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123-11-5
:
Formula:C₈H₈O₂
MDL Number:MFCD00003385
Merck Index:13,00666
Boiling Pt:248…249 °C (760 mmHg)
Melting Pt:–1 °C
Density:1.121 g/cm³
Flash Pt:116 °C
:136.15 g/mol
Storage Temperature:Ambient
EINECS:204-602-6
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5710 to 1.5740 (20°C, 589 nm)
- GC:≥99.0 %
- Relative density:(D20/4°C) 1.120 to 1.124
Specification Test Results
Frequently Bought Together