Order Entry
Czech Republic
ContactUsLinkComponent
4-Hydroxy-3-nitrobenzaldehyde 97%
Irritant
4-Hydroxy-3-nitrobenzaldehyde 97%
Certificates
undefined
Irritant
4-Hydroxy-3-nitrobenzaldehyde 97%

Specification Test Results

  • Appearance:
    Yellow-brown to brown Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    139 to 144 °C
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together