4-(Dimethylamino)benzaldehyde, TECHNICAL
Catalog # 23438.268
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Identification:Passes test
Specification Test Results
About this item
:
:149.19 g/mol
Merck Index:13,03257
Boiling Pt:266 °C (1013 hPa)
Melting Pt:72 °C
Density:1.0254 g/cm³ (99.9 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:202-819-0
CAS Number:100-10-7
Formula:(CH₃)₂NC₆H₄CHO
MDL Number:MFCD00003381