Order Entry
Czech Republic
ContactUsLinkComponent
 
 
2,4-Dibromophenol 99%
  ACRO406510050
 :  
undefined
 
2,4-Dibromophenol 99%
  ACRO406510050
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to light yellow to beige Crystalline powder or crystals or mass and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    36 to 39 °C
  • GC:
    ≥98.5 %

 

 

 :
 :251.91 g/mol
 :154 °C (11 mmHg)
 :36…39 °C
 :2,07 g/cm³ (20 °C)
 :>110 °C
 :210-436-5
 :2811
 :6.1,II
 :GHS06
 :615-58-7
 :C₆H₄Br₂O
 :MFCD00002149