
4-Bromo-1,3-dimethoxybenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17715-69-4
:
Formula:C₈H₉BrO₂
MDL Number:MFCD00009844
:217.06 g/mol
Boiling Pt:153…155 °C (18 mmHg)
Melting Pt:24…26 °C
Density:1.496
Flash Pt:113 °C (235 °F)
EINECS:241-717-0
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together