Methyl-3,4,5-trimethoxybenzoate, Sigma-Aldrich®
Katalogové číslo SIALM86006-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:226.23 g/mol
:M-Clarity™ quality level = MQ100
EINECS:217-629-3
CAS Number:1916-07-0
Formula:C₁₁H₁₄O₅
MDL Number:MFCD00008431