2-Methyl-2-propyl-1,3-propanediol, Sigma-Aldrich®
Catalog # SIALM75209-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:132.2 g/mol
:M-Clarity™ quality level = MQ100
EINECS:201-099-5
CAS Number:78-26-2
Formula:C₇H₁₆O₂
MDL Number:MFCD00004696