Order Entry
Czech Republic
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, 2,4,6-Tris(trifluoromethyl)-1,3,5-triazine
  80286-1G-F
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, 2,4,6-Tris(trifluoromethyl)-1,3,5-triazine
  80286-1G-F
 :  Merck
 :  

 

  • Pk:
    1 g
  • Standards:
    Analytical standard, suitable for mass spectrometry (MS)
  • CAS No.:
    368-66-1
  • Description:
    2,4,6-Tris(trifluoromethyl)-1,3,5-triazine
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302 + H312 + H332 - H319 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00042436
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    285,07
  • Flash point:
    113 °C - Closed cup
  • Boiling point:
    98,3 - 98,5 °C/748 mmHg (lit.)

 

 

  • GHS signal: GHS07
 : P261 - P280 - P301 + P312 - P302 + P352 + P312 - P304 + P340 + P312 - P305 + P351 + P338