
2-Methoxy-1-methylethyl acetate (PGMEA), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-65-6
:
Formula:C₆H₁₂O₃
MDL Number:MFCD00038500
:132.16 g/mol
Boiling Pt:145.8 °C (1013 hPa)
Melting Pt:–67 °C
Density:0.968 g/cm³ (20 °C)
Flash Pt:45 °C
EINECS:203-603-9
UN:3272
ADR:3,III
:GHS02
Frequently Bought Together