2-Hydroxy-5-(trifluoromethyl)pyridine, Sigma-Aldrich®
Catalog # SIAL442801-5G-A
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:163.1 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:33252-63-0
Formula:C₆H₄F₃NO
MDL Number:MFCD00042315