4-(Trifluoromethyl)phenylhydrazine, Sigma-Aldrich®
Catalog # SIAL412295-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:176.14
:M-Clarity™ quality level = MQ200
CAS Number:368-90-1
Formula:C₇H₇F₃N₂
MDL Number:MFCD00042508