Ammonium trifluoroacetate ≥99.0% (by non aqueous titration) for HPLC, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:3336-58-1
:
Formula:C₂H₄F₃NO₂
MDL Number:MFCD00012615
:131.05 g/mol
Melting Pt:123…125 °C
Storage Temperature:Ambient
EINECS:222-072-4
Specification Test Results
- Assay:99.0 - 101.0 %
- Identity (IR):complying
- pH (10 %, 20°C):5 - 6.5
- Colour of the solution:complying
- Turbidity of solution:complying
- Suitability test (HPLC):complying
- Absorbance at 500 nm:Max. 0.02
- Absorbance at 290 nm:Max. 0.02
- Absorbance at 280 nm:Max. 0.03
- Absorbance at 270 nm:Max. 0.04
- Absorbance at 260 nm:Max. 0.05
Specification Test Results
Frequently Bought Together