Order Entry
Czech Republic
ContactUsLinkComponent
Oxalic acid 0.05 mol/l (0.1 N) volumetric solution
Oxalic acid 0.05 mol/l (0.1 N) volumetric solution
Catalog # REGEOA20051
Supplier:  REAGECON
CAS Number:  
Oxalic acid 0.05 mol/l (0.1 N) volumetric solution
Catalog # REGEOA20051
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :90.04 g/mol
Storage Temperature:Ambient
EINECS:205-634-3
ADR:8,III
CAS Number:144-62-7
Formula:C₂H₂O₄
MDL Number:MFCD00002573