Ethylammonium chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:557-66-4
:
Formula:C₂H₈ClN
MDL Number:MFCD00012885
:81.54 g/mol
:M-Clarity™ quality level = MQ200
EINECS:209-182-8
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:107 - 108 °C
- Molar Mass:81.54 g/mol
Specification Test Results
Frequently Bought Together