Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, 2-Methylnaphthalene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Organic reference standard, 2-Methylnaphthalene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20895200CY
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 2-Methylnaphthalene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20895200CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    91-57-6
  • Description:
    2-Methylnaphthalene 10 µg/ml in cyclohexane
  • Formula:
    C₁₁H₁₀
  • Description:
    2-Methylnaphthalene

Specifications

About this item