Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, 1,2,3-Trichloropropane 10 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, 1,2,3-Trichloropropane 10 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERL17780000ME
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 1,2,3-Trichloropropane 10 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERL17780000ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    96-18-4
  • Description:
    1,2,3-Trichloropropane 10 µg/ml in methanol
  • Formula:
    C₃H₅Cl₃
  • Description:
    1,2,3-Trichloropropane

Specifications

About this item