Order Entry
Czech Republic
ContactUsLinkComponent
2,4-Toluenediisocyanate
2,4-Toluenediisocyanate
Catalog # EHERCA17594500
Supplier:  EHRENSTORFER
CAS Number:  
2,4-Toluenediisocyanate
Catalog # EHERCA17594500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :174.16
EINECS:209-544-5
UN:2078
ADR:6.1,II
CAS Number:584-84-9
 :26471-62-5
Formula:C₉H₆N₂O₂
MDL Number:MFCD00002011