Order Entry
Czech Republic
ContactUsLinkComponent
2-Hydroxy-5-(trifluoromethyl)pyridine
2-Hydroxy-5-(trifluoromethyl)pyridine
Catalog # EHERC14253000
Supplier:  EHRENSTORFER
CAS Number:  
2-Hydroxy-5-(trifluoromethyl)pyridine
Catalog # EHERC14253000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :163.1 g/mol
CAS Number:33252-63-0
Formula:C₆H₄F₃NO
MDL Number:MFCD00042315