Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Organic reference standard, Fluoranthene 200 µg/ml in isooctane, CIL
Organic reference standard, Fluoranthene 200 µg/ml in isooctane, CIL
Organic reference standard, Fluoranthene 200 µg/ml in isooctane, CIL
PROOCILULM74211.2
:
LGC Standards PROMOCHEM
:
Organic reference standard, Fluoranthene 200 µg/ml in isooctane, CIL
PROOCILULM74211.2
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle
Pk:
1,2 mL
CAS No.:
206-44-0
Description:
Fluoranthene 200 µg/ml in isooctane
Formula:
C₁₆H₁₀
Description:
Fluoranthene