Order Entry
Czech Republic
ContactUsLinkComponent
Citric acid 50% in aqueous solution, Biotech Reagent, J.T.Baker®
Corrosive
Citric acid 50% in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Citric acid 50% in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
    48 - 52%
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Assay (HOC(COOH)(CH₂COOH)₂):
    48 - 52 %
  • Identification:
    Passes Test
  • Chloride (Cl):
    ≤0.001 %
  • Heavy Metals (as Pb):
    ≤0.001 %
  • ACS - Iron (Fe):
    ≤3 ppm
  • Phosphate (PO₄):
    ≤0.001 %

Specification Test Results

Related Information

Frequently Bought Together