4-Bromobenzaldehyde diethyl acetal ≥98%
Catalog # B25163.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
About this item
CAS Number:34421-94-8
:
Formula:C₁₁H₁₅BrO₂
MDL Number:MFCD01863514
:259.14 g/mol
Boiling Pt:99…101 °C (1 mmHg)
Density:1.274
Flash Pt:104 °C (219 °F)
Storage Temperature:Ambient