
2,4,6-Tris(trifluoromethyl)-1,3,5-triazine ≥98%
Catalog # B22001.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
About this item
:
:285.07 g/mol
Boiling Pt:95…96 °C
Density:1.596
EINECS:206-709-3
:GHS07
Storage Temperature:Ambient
CAS Number:368-66-1
Formula:C₆F₉N₃
MDL Number:MFCD00042436