
2,3-Dimethylphenylhydrazinium hydrochloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56737-75-8
:123333-92-6
Formula:C₈H₁₃ClN₂
MDL Number:MFCD00013380
:172.66 g/mol
Melting Pt:∼ 210 °C (dec.)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together