
4-Chlorophenylhydrazinium chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1073-70-7
:
Formula:C₆H₈Cl₂N₂
MDL Number:MFCD00012943
:179.05 g/mol
Melting Pt:∼ 216 °C (dec.)
EINECS:214-030-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together