
Methylammonium chloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:593-51-1
:
Formula:CH₆ClN
MDL Number:MFCD00012849
:67.52 g/mol
Merck Index:14,6014
Boiling Pt:225…230 °C (15 mmHg)
Melting Pt:230…234 °C
Storage Temperature:Ambient
EINECS:209-795-0
:GHS07
Related Information
Frequently Bought Together